ENA372573043 Enamine

5-[(4-hydroxyphenyl)methyl]oxolan-2-one

  • MDL number MFCD22392160
  • Cas Number : 871329-30-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112078

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112078
Availability details for
Quantity
  • Availability
  • Add to Cart