ENA372573048 Enamine

5-chloro-6-hydrazinylpyridazin-4-amine

  • MDL number MFCD19201393
  • Cas Number : 61071-29-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108601

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108601
Availability details for
Quantity
  • Availability
  • Add to Cart