ENA372573049 Enamine

3-benzyl-3-azabicyclo[3.1.0]hexan-6-amine dihydrochloride

  • MDL number MFCD22392162
  • Cas Number : 1394662-71-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 214...216
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112097

Safety Information

msds https://www.enaminestore.com/catalog/EN300-112097
Availability details for
Quantity
  • Availability
  • Add to Cart