ENA372573052 Enamine

3-amino-1-(pyridin-2-yl)thiourea

  • MDL number MFCD00856418
  • Cas Number : 36273-89-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112100

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112100
Availability details for
Quantity
  • Availability
  • Add to Cart