ENA372573057 Enamine

3,4,5-trifluoro-N'-hydroxybenzene-1-carboximidamide

  • MDL number MFCD07776441
  • Cas Number : 885954-61-0
  • Molecular Weight : 190.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-112106

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112106
Availability details for
Quantity
  • Availability
  • Add to Cart