ENA372573064 Enamine

5-(1-chloroethyl)-3-(4-methoxyphenyl)-1,2,4-oxadiazole

  • MDL number MFCD12189041
  • Cas Number : 91066-38-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108810

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108810
Availability details for
Quantity
  • Availability
  • Add to Cart