ENA372573066 Enamine

5-(chloromethyl)-3-(3,4-difluorophenyl)-1,2,4-oxadiazole

  • MDL number MFCD12110807
  • Cas Number : 1153429-79-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108812

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108812
Availability details for
Quantity
  • Availability
  • Add to Cart