ENA372573076 Enamine

2-{4-[(4-chlorophenyl)methoxy]phenyl}acetic acid

  • MDL number MFCD06208464
  • Cas Number : 125721-51-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112177

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112177
Availability details for
Quantity
  • Availability
  • Add to Cart