ENA372573080 Enamine

2-chloro-N-[2-(2-chlorophenyl)-2-(pyrrolidin-1-yl)ethyl]acetamide hydrochloride

  • MDL number MFCD09277628
  • Cas Number : 1052545-91-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-112183

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112183
Availability details for
Quantity
  • Availability
  • Add to Cart