ENA372573087 Enamine

2-(2-phenyl-1H-imidazol-1-yl)propanoic acid

  • MDL number MFCD09908338
  • Cas Number : 1248743-01-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112196

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112196
Availability details for
Quantity
  • Availability
  • Add to Cart