ENA372573095 Enamine

2-(3,4-difluorophenyl)ethan-1-ol

  • MDL number MFCD00143061
  • Cas Number : 286440-92-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-112205

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112205
Availability details for
Quantity
  • Availability
  • Add to Cart