ENA372573101 Enamine

2-chloro-5-acetamidobenzoic acid

  • MDL number MFCD00466845
  • Cas Number : 719282-11-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112213

Safety Information

msds https://www.enaminestore.com/catalog/EN300-112213
Availability details for
Quantity
  • Availability
  • Add to Cart