ENA372573104 Enamine

3-chloro-5-cyanopyridine-2-carboxylic acid

  • MDL number MFCD14584817
  • Cas Number : 1200497-81-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112219

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112219
Availability details for
Quantity
  • Availability
  • Add to Cart