ENA372573118 Enamine

2-(2-nitrophenyl)ethane-1-sulfonyl chloride

  • MDL number MFCD17285340
  • Cas Number : 3192-07-2
  • Molecular Weight : 249.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112252

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112252
Availability details for
Quantity
  • Availability
  • Add to Cart