ENA372573121 Enamine

5-methylpyrimidine-4-carbonitrile

  • MDL number MFCD18820938
  • Cas Number : 114969-63-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102246

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102246
Availability details for
Quantity
  • Availability
  • Add to Cart