ENA372573124 Enamine

methyl 2-(6-hydroxy-2-methylpyrimidin-4-yl)acetate

  • MDL number MFCD03411899
  • Cas Number : 93715-53-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112261

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112261
Availability details for
Quantity
  • Availability
  • Add to Cart