ENA372573136 Enamine

1-(3-methylfuran-2-yl)ethan-1-one

  • MDL number MFCD22392181
  • Cas Number : 13101-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112275

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112275
Availability details for
Quantity
  • Availability
  • Add to Cart