ENA372573137 Enamine

[2-(tert-butoxy)ethyl](methyl)amine

  • MDL number MFCD03453521
  • Cas Number : 1008119-69-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112277

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112277
Availability details for
Quantity
  • Availability
  • Add to Cart