ENA372573140 Enamine

5,5-dimethylcyclopent-1-ene-1-carboxylic acid

  • MDL number MFCD19231111
  • Cas Number : 154222-75-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112280

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112280
Availability details for
Quantity
  • Availability
  • Add to Cart