ENA372573161 Enamine

(4-methyl-4H-1,2,4-triazol-3-yl)methanol

  • MDL number MFCD13190102
  • Cas Number : 59660-30-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 49...51
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112346

Safety Information

msds https://www.enaminestore.com/catalog/EN300-112346
Availability details for
Quantity
  • Availability
  • Add to Cart