ENA372573165 Enamine

5-fluoro-1-benzothiophene-2-sulfonyl chloride

  • MDL number MFCD22199595
  • Cas Number : 128851-97-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105671

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105671
Availability details for
Quantity
  • Availability
  • Add to Cart