ENA372573166 Enamine

2-(piperidin-3-yl)-4,5,6,7-tetrahydro-1,3-benzothiazole

  • MDL number MFCD09891531
  • Cas Number : 1017184-36-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112354

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112354
Availability details for
Quantity
  • Availability
  • Add to Cart