ENA372573169 Enamine

[5-(2-methylphenyl)-1,2,4-oxadiazol-3-yl]methanol

  • MDL number MFCD22392190
  • Cas Number : 1394645-03-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112360

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112360
Availability details for
Quantity
  • Availability
  • Add to Cart