ENA372573179 Enamine

tert-butyl 3-sulfamoylpyrrolidine-1-carboxylate

  • MDL number MFCD22392191
  • Cas Number : 1394669-98-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112410

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112410
Availability details for
Quantity
  • Availability
  • Add to Cart