ENA372573189 Enamine

3-{[4-(trifluoromethoxy)phenyl]methyl}azetidine hydrochloride

  • MDL number MFCD22392195
  • Cas Number : 1394688-94-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112446

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112446
Availability details for
Quantity
  • Availability
  • Add to Cart