ENA372573211 Enamine

(furan-2-yl)(2-methoxyphenyl)methanamine

  • MDL number MFCD16086849
  • Cas Number : 1248791-81-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112498

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112498
Availability details for
Quantity
  • Availability
  • Add to Cart