ENA372573213 Enamine

2-(cyclopropylamino)acetamide

  • MDL number MFCD09814605
  • Cas Number : 1016674-92-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112500

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112500
Availability details for
Quantity
  • Availability
  • Add to Cart