ENA372573218 Enamine

2-(2-bromo-5-methoxyphenyl)acetic acid

  • MDL number MFCD11554152
  • Cas Number : 86826-93-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112506

Safety Information

msds https://www.enaminestore.com/catalog/EN300-112506
Availability details for
Quantity
  • Availability
  • Add to Cart