ENA372573222 Enamine

2-chloro-N-[(4-methylphenyl)methyl]pyrimidin-4-amine

  • MDL number MFCD09756914
  • Cas Number : 945895-76-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112511

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112511
Availability details for
Quantity
  • Availability
  • Add to Cart