ENA372573223 Enamine

2-cyclopropyl-6-methoxyquinoline-4-carboxylic acid

  • MDL number MFCD05176306
  • Cas Number : 897565-71-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112516

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112516
Availability details for
Quantity
  • Availability
  • Add to Cart