ENA372573234 Enamine

2-(3-methoxypropanesulfonamido)acetic acid

  • MDL number MFCD18309952
  • Cas Number : 1341427-02-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112561

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112561
Availability details for
Quantity
  • Availability
  • Add to Cart