ENA372573237 Enamine

1-(4-acetyl-1-methyl-1H-pyrrol-2-yl)-2,2,2-trichloroethan-1-one

  • MDL number MFCD00141901
  • Cas Number : 338753-30-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 133...135
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112566

Safety Information

msds https://www.enaminestore.com/catalog/EN300-112566
Availability details for
Quantity
  • Availability
  • Add to Cart