ENA372573239 Enamine

2-(4-formyl-1H-1,2,3-triazol-1-yl)acetic acid

  • MDL number MFCD20660491
  • Cas Number : 1175146-73-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112568

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112568
Availability details for
Quantity
  • Availability
  • Add to Cart