ENA372573244 Enamine

4-(3,5-dimethyl-1H-pyrazol-1-yl)benzene-1-sulfonic acid

  • MDL number MFCD03216931
  • Cas Number : 91088-11-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112584

Safety Information

msds https://www.enaminestore.com/catalog/EN300-112584
Availability details for
Quantity
  • Availability
  • Add to Cart