ENA372573247 Enamine

4-(2-bromoethyl)-1,2-difluorobenzene

  • MDL number MFCD12025017
  • Cas Number : 175018-77-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112592

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-112592
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart