ENA372573248 Enamine

6-chloro-N,N-dimethylpyridazine-3-carboxamide

  • MDL number MFCD12165825
  • Cas Number : 1183315-30-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 137...139
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105947

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105947
Availability details for
Quantity
  • Availability
  • Add to Cart