ENA372573249 Enamine

4-(tert-butylsulfanyl)benzonitrile

  • MDL number MFCD11136358
  • Cas Number : 132555-38-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112595

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112595
Availability details for
Quantity
  • Availability
  • Add to Cart