ENA372573254 Enamine

2-amino-4-oxo-4-phenylbutanoic acid hydrochloride

  • MDL number MFCD00136889
  • Cas Number : 54631-37-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 208-210
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-112603

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112603
Availability details for
Quantity
  • Availability
  • Add to Cart