ENA372573276 Enamine

2-methyl-6-(morpholin-4-yl)pyridin-3-amine

  • MDL number MFCD11849888
  • Cas Number : 221159-08-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-112673

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112673
Availability details for
Quantity
  • Availability
  • Add to Cart