ENA372573280 Enamine

ethyl 2-methylpyrrolidine-2-carboxylate

  • MDL number MFCD20305101
  • Cas Number : 16277-07-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112677

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112677
Availability details for
Quantity
  • Availability
  • Add to Cart