ENA372573281 Enamine

(1-{[1,2,4]triazolo[4,3-a]pyrazin-8-yl}piperidin-3-yl)methanamine

  • MDL number MFCD20085931
  • Cas Number : 1421601-99-1
  • Molecular Weight : 232.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (1-[1,2,4]triazolo[4,3-a]pyrazin-8-yl-3-piperidinyl)methanamine
Inchi Code 1S/C11H16N6/c12-6-9-2-1-4-16(7-9)10-11-15-14-8-17(11)5-3-13-10/h3,5,8-9H,1-2,4,6-7,12H2
Inchi Key RSAZSAAIDOOJKY-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-112678

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112678
Availability details for
Quantity
  • Availability
  • Add to Cart