ENA372573282 Enamine

3-(2,6-difluorophenyl)aniline

  • MDL number MFCD19637699
  • Cas Number : 851462-21-0
  • Molecular Weight : 205.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-112680

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112680
Availability details for
Quantity
  • Availability
  • Add to Cart