ENA372573292 Enamine

5-(2,4-dimethylphenyl)-5-methylimidazolidine-2,4-dione

  • MDL number MFCD09736307
  • Cas Number : 1052563-94-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144...146
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109151

Safety Information

msds https://www.enaminestore.com/catalog/EN300-109151
Availability details for
Quantity
  • Availability
  • Add to Cart