ENA372573297 Enamine

4-[1-(4-methylbenzenesulfonyl)piperidin-4-yl]-1,3-thiazol-2-amine

  • MDL number MFCD09042545
  • Cas Number : 926240-09-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112726

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112726
Availability details for
Quantity
  • Availability
  • Add to Cart