ENA372573321 Enamine

2-methoxypropan-1-ol

  • MDL number MFCD00126948
  • Cas Number : 1589-47-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-112785

Safety Information

signalword Danger
pcodes P210,P233,P240,P241,P242,P243,P261,P271,P280,P303+P361+P353,P304+P340,P312,P370+P378,P403+P233,P403+P235,P405,P501
hcodes F
MSDS https://www.enaminestore.com/catalog/EN300-112785
pictograms GHS02,GHS07
hstatements H225,H336
Availability details for
Quantity
  • Availability
  • Add to Cart