ENA372573330 Enamine

methyl 3-amino-4-[(5-chloropyridin-2-yl)sulfanyl]benzoate

  • MDL number MFCD20561681
  • Cas Number : 1379272-52-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112797

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112797
Availability details for
Quantity
  • Availability
  • Add to Cart