ENA372573331 Enamine

{imidazo[1,2-a]pyridin-5-yl}methanol

  • MDL number MFCD13183181
  • Cas Number : 167884-17-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112798

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112798
Availability details for
Quantity
  • Availability
  • Add to Cart