ENA372573342 Enamine

(4-propyl-4H-1,2,4-triazol-3-yl)methanol

  • MDL number MFCD18819895
  • Cas Number : 497855-00-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112811

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112811
Availability details for
Quantity
  • Availability
  • Add to Cart