ENA372573345 Enamine

2-(cyclopentyloxy)-4-fluoroaniline

  • MDL number MFCD09730450
  • Cas Number : 953740-21-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112815

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112815
Availability details for
Quantity
  • Availability
  • Add to Cart