ENA372573346 Enamine

4-bromo-5-(chlorosulfonyl)-2-fluorobenzoic acid

  • MDL number MFCD11650163
  • Cas Number : 1099186-78-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147...149
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112816

Safety Information

msds https://www.enaminestore.com/catalog/EN300-112816
Availability details for
Quantity
  • Availability
  • Add to Cart